ContactUsLinkComponent
EPA method 501 Trihalomethane Mixture, Restek
EPA method 501 Trihalomethane Mixture, Restek
Catalog # RK30036
Supplier:  Restek
CAS Number:  
undefined
EPA method 501 Trihalomethane Mixture, Restek
Catalog # RK30036
Supplier:  Restek
Supplier Number:  30036
CAS Number:  
Specifications
  • Description:
    EPA Method 501 Trihalomethane Mixture
  • Cat. No.:
    RK30036
  • Supplier no.:
    30036
Specifications
About this item
Mixture is packaged in purge and trap methanol at a concentration of 200 µg/mL in a 1 ml ampoule.
  • Mixture contains bromodichloromethane, bromoform, chloroform, and dibromochloromethane
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).