Nalgene® Bio-Safe Carrier, Polycarbonate, Thermo Scientific
Catalog # 56609-112
Supplier: Thermo Fisher Scientific
Specifications
- Description:Carrier
- Dimensions:37L x 18.4H x 17W cm (141/2 x 71/4 x 63/4")
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. No.:56609-112
- Supplier no.:7135-0001
Specifications
About this item
This closed-system carrier is designed to help protect user and hazardous samples during transportation. Polycarbonate carrier is tough and break-resistant. It offers excellent visibility of contents. Easy-to-grasp side handles are molded in. Clamps securely hold carrier closed and ensure leakproof seal. Carrier is suitable for use with NALGENE® Unwire™ test tube racks with tube O.D. of 13 or 16 mm (see 66023-520, 66023-663, and 60914-605 series).
- Closed-system carrier protects user and hazardous samples during transportation
- Tough, break-resistant polycarbonate with excellent visibility
- Easy-to-grasp side handles are molded in
- Clamps securely hold carrier closed and ensure leakproof seal
- Suitable for use with NALGENE® Unwire™ test tube racks with tube O.D. of 13 or 16 mm
Optional stainless steel handle 56609-111 eases one-handed carrying.
Note: To avoid exposure to biohazardous material in the event of accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.