ContactUsLinkComponent
Spherical Pinch Clamps, Kemtech America
Catalog # 60004-972
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Spherical Pinch Clamps, Kemtech America
Catalog # 60004-972
Supplier:  Kemtech America
Supplier Number:  CC0012
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. No.:
    60004-972
  • Joint size:
    12/5
  • Supplier no.:
    CC0012
  • O-Ring joint size:
    5
Specifications
About this item
Kemtech pinch clamp for spherical joints.
  • Stainless steel construction
  • Adjustable metal tensioner
For use with 12/5 ball joint, or #5 O-ring joints.