N Series Fume Extractor Fans and Manual Starters, Nederman
Supplier: Nederman
Certificates
About this item
Designed for Nederman extractor arms
The simple and powerful fans extract noxious fumes and vapors for various applications. Single or three-phase 60Hz fans are available. The fan starters feature a lockable on/off switch and can be used for both single and three-phase fans. Fans and fan starters are also equipped with motor overload protectors.
Frequently Bought Together