Accessories for Synthware Flasks with High Vacuum Valve
Supplier: Kemtech America
Certificates
About this item
Kemtech Teflon® protected high vacuum valve.
- Teflon® protected O-ring
- For externally threaded glassware
- Teflon® construction
- 0 to 8mm or 0 to 12 mm
This design provides for an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.
Frequently Bought Together