Desoldering Tips for PACE®, Air-Vac™, and A.P.E.™ Desoldering Irons, Plato™, ITW Chemtronics
Supplier: ITW Chemtronics
Certificates
About this item
Made of oxygen-free, high-conductivity copper, these tips are protected inside and outside to minimize erosion by solder.
Tips 15551-306 and -308 are ideal for PACE® SX-20, SX-25, and SX-25V Sodr-X-Tractor™ desoldering handpieces. Tip 15551-312 is ideal for PACE® SX-40A, SX-55A, SX-65A, and SX-70 Sodr-X-Tractor™ desoldering handpieces.
Frequently Bought Together