Order Entry
United States
ContactUsLinkComponent
StatSpin® Cytospin CytoFuge® 2 Compact Cytocentrifuge System, Iris Sample Processing
StatSpin® Cytospin CytoFuge® 2 Compact Cytocentrifuge System, Iris Sample Processing
Catalog # 87002-196
Supplier:  Avantor
UL Listed
undefined
StatSpin® Cytospin CytoFuge® 2 Compact Cytocentrifuge System, Iris Sample Processing
Catalog # 87002-196
Supplier:  Avantor
UL Listed

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Max. Capacity:
    50–800 µL
  • Max. Speed:
    600 - 4400 rpm
  • Description:
    Cytocentrifuge
  • Electrical:
    100–240V, 50/60Hz
  • Max. RCF:
    20–1060 ×g
  • Acceleration/Braking Time:
    20 s (to 95% of speed) / <30 second s
  • Cycle Time:
    2, 4, 6, 8, 10, 30 min
  • Dimensions:
    13.5H x 16.8W x 21.8D cm (59/32 x 619/32 x 819/32")
  • Weight:
    13.2 kg (6 lbs.)
  • Cat. No.:
    87002-196
  • Supplier no.:
    CF02

Specifications

About this item

With simple controls and snap-seal filter concentrators for easy operation, this economical centrifuge is the perfect solution for your cytocentrifuge needs.

  • Efficient and Quiet Processing
  • Consistent Results
  • Simple Controls
  • Convenient Size
  • Snap-Seal Filter Concentrators

It processes samples quickly, silently, and conveniently on any bench or in a safety cabinet.

High-quality slide preps with simple, standardized procedures.

Certifications: UL listed. CE marked. Meets standards for mechanical, electrical, and biohazard safety.
Ordering information: Includes manufacturer’s one-year warranty, four-place rotor with lid, four disposable one-well cell concentrators, four disposable three-well cell concentrators, eight clips, 14 backing plates, eight disposable filter concentrators, four replacement clips for disposable filter concentrator, universal power supply, and 120V adapter with power cord.