Revco® Digital Temperature Monitors, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These digital temperature and power monitor units provide both alarm/monitoring and digital temperature display in a single, self-contained device.
- An audible and visual warning is activated in case of power failure or overtemperature condition
- Monitors can be installed on or adjacent to any freezer
One channel monitors feature a Velcro closure, 2 or 3 channel monitors feature a mounting bracket. Standard features include high alarm with mute and chirp, audio delay timer, remote alarm contacts (SPDT), LCD numeric display, NiCad battery backup, low voltage AC power supply (120 V), and 32L×6D mm (1¹/₄×¹/₄") temperature probe. The 2 or 3 channel monitors are supplied with a high and low alarm, and 8-character backlit adjustable LCD alphanumeric display.
Specifications
- Catalog No:
- 14228-506
- 14228-502
- 14228-504
- Alarm:
- High and low temperature alarm, visual alarm (green, red)
- High temperature alarm, red visual alarm
- High temperature alarm, red visual alarm
- Display:
- 8-character, back-lit, adjustable LCD alphanumeric display
- Numeric-only LCD alphanumeric display
- Numeric-only LCD alphanumeric display
- W×D×H:
- 83×50×158 mm
- 114×50×27 mm
- 114×50×27 mm