Orders LinkContactUsLinkComponent
Potter-Elvehjem Tissue Grinders, Wheaton
Potter-Elvehjem Tissue Grinders, Wheaton
Potter-Elvehjem Tissue Grinders, Wheaton
Certificates
Potter-Elvehjem Tissue Grinders, Wheaton
Potter-Elvehjem Tissue Grinders, Wheaton
19 Options Available Below
Product Details & Documents
Grinders for preparation of soft tissue homogenates, such as liver, brain.
  • Grinders are available with smooth pestles or serrated pestles, which disperse homogenate into the mortar cylinder more effectively
  • Depth of radial serrations is 0.508–0.635 mm (0.020–0.025")
  • Smooth or serrated pestles are available
  • Autoclavable and parts are interchangeable
All pestles are made of fluoropolymer resin with a 6.4 mm (1/4") diameter stainless steel shaft. Radius at pestle bottom conforms to radius in bottom of grinding chamber. Grinding chamber is made of low extractable borosilicate glass. Chamber clearance is 0.102–0.152 mm (0.004–0.006").
Certifications: Glass meets ASTM Type 1 Class A and USP Type 1 standards.
Product Family Options
Product Information
  • Capacity
    Overall length
    Chamber dimensions
    Availability
    Price
  • 14231-368
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    15 ml
    219 mm
    19O.D. x 84L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $385.90
    $385.90
    New Customer Promotional Price
    Case of 2
       $385.90
    Minimum order quantity is 1
    Supplier Number: 357984
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-368
    Small Business Enterprise
  • 14231-370
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    30 ml
    266 mm
    24O.D. x 118L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $443.70
    $443.70
    New Customer Promotional Price
    Case of 2
       $443.70
    Minimum order quantity is 1
    Supplier Number: 357989
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-370
    Small Business Enterprise
  • 14231-372
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    55 ml
    266 mm
    30O.D. x 130L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $523.60
    $523.60
    New Customer Promotional Price
    Case of 2
       $523.60
    Minimum order quantity is 1
    Supplier Number: 357994
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-372
    Small Business Enterprise
  • 14231-384
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    10 ml
    219 mm
    16O.D. x 74L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $340.85
    $340.85
    New Customer Promotional Price
    Case of 2
       $340.85
    Minimum order quantity is 1
    Supplier Number: 357979
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-384
    Small Business Enterprise
  • 14231-340
    Grinding Chamber Only
    Grinding Chamber Only
    2 ml
    11O.D. x 45L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $158.10
    $158.10
    New Customer Promotional Price
    Case of 2
       $158.10
    Minimum order quantity is 1
    Supplier Number: 358028
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-340
    Small Business Enterprise
  • 14231-342
    Grinding Chamber Only
    Grinding Chamber Only
    5 ml
    13O.D. x 66L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $151.30
    $151.30
    New Customer Promotional Price
    Case of 2
       $151.30
    Minimum order quantity is 1
    Supplier Number: 358033
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-342
    Small Business Enterprise
  • 14231-364
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    2 ml
    203 mm
    11O.D. x 45L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $331.50
    $331.50
    New Customer Promotional Price
    Case of 2
       $331.50
    Minimum order quantity is 1
    Supplier Number: 357969
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-364
    Small Business Enterprise
  • 14231-366
    Grinding Chamber and Serrated Pestle
    Grinding Chamber and Serrated Pestle
    5 ml
    219 mm
    13O.D. x 66L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $335.75
    $335.75
    New Customer Promotional Price
    Case of 2
       $335.75
    Minimum order quantity is 1
    Supplier Number: 357974
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-366
    Small Business Enterprise
  • 14231-374
    Grinding Chamber Only
    Grinding Chamber Only
    15 ml
    19O.D. x 84L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $167.45
    $167.45
    New Customer Promotional Price
    Case of 2
       $167.45
    Minimum order quantity is 1
    Supplier Number: 358043
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-374
    Small Business Enterprise
  • 14231-376
    Grinding Chamber Only
    Grinding Chamber Only
    55 ml
    30O.D. x 130L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $237.15
    $237.15
    New Customer Promotional Price
    Case of 2
       $237.15
    Minimum order quantity is 1
    Supplier Number: 358053
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14231-376
    Small Business Enterprise
  • 62400-700
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    2 ml
    203 mm
    11O.D. x 45L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $162.35
    $162.35
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $324.70
    $324.70
    New Customer Promotional Price
    Each
       $162.35
    Case of 2
       $324.70
    Supplier Number: 358029
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-700
    Small Business Enterprise
  • 62400-722
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    5 ml
    219 mm
    13O.D. x 66L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $164.90
    $164.90
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $329.80
    $329.80
    New Customer Promotional Price
    Each
       $164.90
    Case of 2
       $329.80
    Supplier Number: 358034
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-722
    Small Business Enterprise
  • 62400-733
    Grinding Chamber Only
    Grinding Chamber Only
    10 ml
    219 mm
    16O.D. x 74L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $81.60
    $81.60
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $163.20
    $163.20
    New Customer Promotional Price
    Each
       $81.60
    Case of 2
       $163.20
    Supplier Number: 358038
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-733
    Small Business Enterprise
  • 62400-744
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    10 ml
    219 mm
    16O.D. x 74L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $166.60
    $166.60
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $333.20
    $333.20
    New Customer Promotional Price
    Each
       $166.60
    Case of 2
       $333.20
    Supplier Number: 358039
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-744
    Small Business Enterprise
  • 62400-755
    Smooth Pestle Only
    Smooth Pestle Only
    10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $86.70
    $86.70
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $173.40
    $173.40
    New Customer Promotional Price
    Each
       $86.70
    Case of 2
       $173.40
    Supplier Number: 358036
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-755
    Small Business Enterprise
  • 62400-766
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    15 ml
    219 mm
    19O.D. x 84L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $190.40
    $190.40
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $380.80
    $380.80
    New Customer Promotional Price
    Each
       $190.40
    Case of 2
       $380.80
    Supplier Number: 358044
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-766
    Small Business Enterprise
  • 62400-777
    Grinding Chamber Only
    Grinding Chamber Only
    30 ml
    266 mm
    24O.D. x 118L mm
    Discontinued
    Supplier Number: -
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-777
    Small Business Enterprise
  • 62400-788
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    30 ml
    266 mm
    24O.D. x 118L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $221.85
    $221.85
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $443.70
    $443.70
    New Customer Promotional Price
    Each
       $221.85
    Case of 2
       $443.70
    Supplier Number: 358049
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-788
    Small Business Enterprise
  • 62400-799
    Smooth Pestle Only
    Smooth Pestle Only
    30 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-799
    Small Business Enterprise
  • 62400-802
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    55 ml
    266 mm
    30O.D. x 130L mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $263.50
    $263.50
    New Customer Promotional Price
    Log in for availability
    Case of 2
    Case of 2 - $527.00
    $527.00
    New Customer Promotional Price
    Each
       $263.50
    Case of 2
       $527.00
    Supplier Number: 358054
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-802
    Small Business Enterprise
  • 62400-879
    Grinding Chamber and Smooth Pestle
    Grinding Chamber and Smooth Pestle
    0.1 ml
    110 mm
    4O.D x 65L mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $198.90
    $198.90
    New Customer Promotional Price
    Case of 2
       $198.90
    Minimum order quantity is 1
    Supplier Number: 357844
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    62400-879
    Small Business Enterprise