Dry Blending Container, Waring
Dry Blending Container, Waring®
Catalog # 58983-100
Supplier: Waring Products
Supplier #: SS115
Jump to:
Product Details & Documents
Unique blade configuration designed for pulverizing and homogenizing dry substances
Note: Blending such as soil processing requires the use of a larger motor unit.
One liter stainless steel container and lid. Fits 1L Waring® blenders.
Specifications
- Description:Dry Blending Container, Waring®