Whatman™ VACU-GUARD Disk Filters, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
Whatman VACU-GUARD disk filters are ideal for protecting vacuum pump systems from aqueous aerosols and water intrusion.
- Designed for in-line use with stepped barb connections
- Membrane retains 99.99% of airbone particles greater than 0.1 µm
- Features hydrophobic polytetrafluoroethylene (PTFE) membrane
Available in 50 and 60 mm formats.
Ordering information: All materials pass USP Class VI test for plastics.
Specifications
- Catalog No:
- 28137-737
- 28137-858
- Application:
- Vacuum pump protection.
Activated carbon removes organic vapors and radioactive particles.
Molecular sieve for use with organic and alkaline air streams.
Desiccant for use with high velocity acidic air streams.
Removes a potential health hazard from the work place. - Vacuum pump protection.
Activated carbon removes organic vapors and radioactive particles.
Molecular sieve for use with organic and alkaline air streams.
Desiccant for use with high velocity acidic air streams.
Removes a potential health hazard from the work place.
- Housing Material:
- Polypropylene (PP)
- Polypropylene (PP)
- Max. Operating Pressure:
- 1 bar (15 psi)
- 1 bar (15 psi)
- Retention:
- Aqueous solutions: up to 0.9 bar (14 psi)
Particulates in air: 0.1 µm 99.99% - Aqueous solutions: up to 0.9 bar (14 psi)
Particulates in air: 0.1 µm 99.99%
Specifications
Related Information
Certificates
Search by Lot Number28420219_DATA FILE
Datasheet
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together