Whatman™ Grade 8 Ruled Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
These white cotton linter cellulose filters are ideal for routine investigations of foreign substances in a variety of sample types.
- Resistant to aqueous and non aqueous solutions
Printed green lines for optical assessment at 5 mm intervals.
Specifications
- Catalog No:
- 28444-045
- 28444-023
- 28444-067
- 10035-654
- Basis Weight:
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together