ContactUsLinkComponent
Multi-Well Plate Vacuum Manifold, Cytiva (Formerly Pall Lab)
  16003-836
 :  Cytiva
Product available on GSA Advantage®
undefined
Multi-Well Plate Vacuum Manifold, Cytiva (Formerly Pall Lab)
  16003-836
 :  Cytiva
 :  5017
Product available on GSA Advantage®
  • Description:
    Multi-Well Plate Vacuum Manifold
  • Environmentally Preferable:
  • Cat. No.:
    16003-836
  • Supplier no.:
    5017
Optimized for the vacuum filtration of AcroPrep™ and AcroWell™ multi-well plates, manifold can also be used with other multi-well filter plates that meet ANSI/SBS X-2004 specifications.
  • For vacuum filtration
  • Can be used with other multi-well plates
  • Anodized aluminum
  • Adapter collar holds filter plates tightly to receiver plates for centrifugation
Manifold is constructed of anodized aluminum and is designed to facilitate the filtration of liquid through a multi-well filter plate into a receiver plate for collection.
Vacuum manifold unit includes a Delrins spacer block designed to accommodate standard 350 μL receiver plates. The spacer block has been optimized to reduce the space between the receiver plate and the filter plate during vacuum filtration. Optional spacer block available for use with 1 mL receiver plates.
 : Manifold includes O-ring, gasket, and spacer block. Control block on manifold includes vacuum pressure gauge, metering valve, release valve, and 6.4mm (1/4") hose barb connection for vacuum line attachment. Replacement accessory kit includes O-ring, gasket, and allen wrench.