VWR® Classical Liquid Media, Dulbeccos Modified Eagles Medium, High Glucose
About this item
VWR® DMEM high glucose liquid media supports cell growth in research, upstream process development, and cell culture manufacturing processes.
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
DMEM media contain increased concentrations of vitamins, minerals, and amino acids compared to traditional basal media. DMEM medium composition also includes ferric nitrate, making this cell culture medium an option for certain cell lines requiring iron for cell growth. DMEM media preparations are available in high or low glucose formulations, as well as with or without L-glutamine, magnesium, and sodium pyruvate. DMEM formulation options may also include phenol red as a pH indicator. All available DMEM media products contain a sodium bicarbonate buffer with or without HEPES to maintain optimal culture pH.
VWR® DMEM raw material components are screened through strict quality control testing for lot-to-lot consistency. DMEM liquid media products are hydrated using purified water and undergo sterile filtration.
DMEM is used for a variety of mammalian cell culture manufacturing processes and research applications. DMEM media formulations have supported tumorigenicity studies, virus replication analyses, and investigations into the influence of cell culture media on gene expression. DMEM medium is commonly used for adherent cells and has been the cell culture medium of choice for hybridoma, fibroblast, neuron, and smooth muscle cell lines as well as many other tissues types. DMEM medium is frequently combined with fetal bovine serum (FBS) as DMEM media composition does not include the growth factors, proteins, hormones, or lipids needed to promote cell growth.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
VWR® DMEM high glucose liquid media supports cell growth in research, upstream process development, and cell culture manufacturing processes.
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
DMEM media contain increased concentrations of vitamins, minerals, and amino acids compared to traditional basal media. DMEM medium composition also includes ferric nitrate, making this cell culture medium an option for certain cell lines requiring iron for cell growth. DMEM media preparations are available in high or low glucose formulations, as well as with or without L-glutamine, magnesium, and sodium pyruvate. DMEM formulation options may also include phenol red as a pH indicator. All available DMEM media products contain a sodium bicarbonate buffer with or without HEPES to maintain optimal culture pH.
VWR® DMEM raw material components are screened through strict quality control testing for lot-to-lot consistency. DMEM liquid media products are hydrated using purified water and undergo sterile filtration.
DMEM is used for a variety of mammalian cell culture manufacturing processes and research applications. DMEM media formulations have supported tumorigenicity studies, virus replication analyses, and investigations into the influence of cell culture media on gene expression. DMEM medium is commonly used for adherent cells and has been the cell culture medium of choice for hybridoma, fibroblast, neuron, and smooth muscle cell lines as well as many other tissues types. DMEM medium is frequently combined with fetal bovine serum (FBS) as DMEM media composition does not include the growth factors, proteins, hormones, or lipids needed to promote cell growth.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77661-374
- 77661-354
- 77661-364
- 77661-366
- 77661-330
- L-Glutamine:
- No
- Yes
- No
- Yes
- Sodium pyruvate:
- Yes
- No
- No
- Yes
- Storage Temperature:
- 2...8 °C
- 2...8 °C
- 2...8 °C
- 2...8 °C
- 2...8 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 77661-374Without L-Glutamine, with sodium pyruvateItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77661-374L-GlutamineNoSodium pyruvateYesStorage Temperature2...8 °C - 77661-354Without L-Glutamine and sodium pyruvateItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77661-354L-GlutamineYesSodium pyruvateNoStorage Temperature2...8 °C - 77661-364DMEM/high: With 4,500 mg/L glucose, with 4.0 mM L-Glutamine and 110 mg/L sodium pyruvateDMEM/high: With 4,500 mg/L glucose, with 4.0 mM L-Glutamine and 110 mg/L sodium pyruvate500 mlLog in for availabilityEach(1/Itm/Bx)$13.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77661-364L-GlutamineNoSodium pyruvateNoStorage Temperature2...8 °C - 77661-366DMEM/high: With 4500 mg/L glucose, 4.0 mM L-Glutamine, and 110 mg/L sodium pyruvateDMEM/high: With 4500 mg/L glucose, 4.0 mM L-Glutamine, and 110 mg/L sodium pyruvate500 mlLog in for availabilityEach(1/Itm/Bx)$76.30Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77661-366L-GlutamineSodium pyruvateStorage Temperature2...8 °C - 77661-330DMEM/high: with 4.0 mM L-Glutamine, without sodium pyruvateDMEM/high: with 4.0 mM L-Glutamine, without sodium pyruvate500 mlLog in for availabilityEach(1/Itm/Bx)$14.49Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77661-330L-GlutamineYesSodium pyruvateYesStorage Temperature2...8 °C