VWR® Dulbecco's Phosphate Buffered Saline Solutions
Supplier: VWR International
New Product
Certificates
About this item
Balanced salt solution used for multiple cell culture applications, (liquid form).
For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Caution: Not for diagnostic or therapeutic use.
Frequently Bought Together