Taq 2X Master Mix
Supplier: EMPIRICAL BIOSCIENCE
New Product
Certificates
About this item
Taq 2X MasterMix supplied as a 2X reaction buffer with dCTP, dGTP, dATP, dTTP, MgCl₂, and Taq DNA polymerase. Taq DNA polymerase gene was isolated from thermus aquaticus YT1 and expressed in E. coli. Empirical's taq 2X MasterMix is easy to use, versatile, and provides excellent amplification for many PCR based applications. Just add template and primers with the MasterMix and the reaction is ready to go.
- Endpoint PCR
- cGMP Manufacturing
Certifications: ISO 13485.
Specifications
- Storage Conditions:Long term storage at −20 °C recommended. Minimize number of freeze thaw cycles of frozen product.