Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for Starter Electrodes
Accessories for Starter Electrodes
Accessories for Starter Electrodes
Catalog # 77679-490
Supplier:
Ohaus
New Product
Expand image
Accessories for Starter Electrodes
Catalog # 77679-490
Supplier:
Ohaus
Supplier Number:
30693682
New Product
Specifications
Description:
Maintenance kit
Cat. No.:
77679-490
For use with:
STDO11: DO electrode
Specifications
About this item
Product Details
Product Details