RTScript™ Reverse Transcriptase
Supplier: EMPIRICAL BIOSCIENCE
New Product
Certificates
About this item
RTScript™ cDNA Transcriptase is a Modified version of M-MLV Reverse Transcriptase with RNase H activity deactivated and increased thermal stability. In combination with optimized buffers and reagents, Empirical's RTScript™ enzyme synthesizes complementary DNA from RNA initiated from RNA or DNA Primers.
- Storage and handling: Store at −20 °C upon arrival until provided expiration date. Minimize Freeze Thaw to avoid loss of performance
Empirical's RTScript™ is sensitive, specific, and is capable of synthesizing highly structured and long cDNA fragments. Produced cDNA is appropriate for both PCR and qPCR analysis. RTScript™ cDNA Transcriptase is supplied with 5X RTScript™ Reaction Buffer and 100 mM DTT.
Frequently Bought Together