About this item
This enzyme is a uracil N-glycosylase (UNG) obtained from the bacterium Esherichia coli. UNG enzymes specifically hydrolyze the N-glycosidic bond of uracil residues in ss/dsDNA molecules leaving an abasic site in the strand but will not react with RNA. The optimal temperature range for E. coli UNG activity is from 30 to 40 °C. The enzyme is irreversibly inactivated after ten minutes of incubation at temperatures ≥95 °C making it suited for carryover prevention in standard PCR and qPCR applications. It is active in most standard reaction buffers used for derivatives of Taq and Pfu DNA polymerases.
Applications: Carryover prevention
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:E. coli
- Enzyme Name:Uracil N-glycosylase