ActiTemp-50 RTScript™ Reverse Transcriptase Kits
Supplier: EMPIRICAL BIOSCIENCE
New Product
Certificates
About this item
Empirical's ActiTemp-50 RTScript™ Reverse Transcriptase is a modified version of M-MLV Reverse Transcriptase with RNase H activity deactivated and increased thermal stability. The enzyme is Warm-Start modified with a proprietary RNA Aptamer.
- Store at −20 °C upon arrival
Activation of the enzyme is achieved instantly by heating to 55 °C to carryout first-strand cDNA synthesis, and results in better specificity compared to unmodified RT. In combination with optimized buffers and reagents, Empirical's ActiTemp-50 RTScript™ enzyme synthesizes complementary DNA from RNA initiated from RNA or DNA Primers.
Applications: Fast Reverse Transcription PCR/qPCR, Inhibitor Tolerant Reverse Transcription PCR/qPCR, Reverse Transcription PCR/qPCR
Certifications: ISO 13485 certified
Frequently Bought Together