HIV-1 Recombinant gp41L (fromE. coli)
Catalog # 77682-614
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV-1
- Protein Synonyms:gp41|gp41L immunodominant regions|gp41 immunodominant regions
- Protein/Peptide Name:gp41L
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-page
- Endotoxin Level:Very low
- Formulation:8 M Urea, 20 mM tris-HCl pH 8.0, 10 mM b-mercaptoethanol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-614
Specifications
About this item
Recombinant HIV-1 gp41 Long Antigen, b-galactosidase-tagged.