HIV Recombinant gp41 (fromE. coli)
77682-596
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV
- Protein Synonyms:gp41 immunodominant region
- Protein/Peptide Name:gp41
- Purity:Greater than 95.0% as determined by SDS-page
- Molecular Weight:13 kDa
- Endotoxin Level:Very low
- Formulation:Lyophilized from 1 mg/ml in 20 mM Na-carbonate, pH 9.6
- Shipping Temperature:Dry Ice
- Cat. No.:77682-596
Recombinant HIV Type-O gp41 Antigen, His-tagged.