HIV Recombinant gp41 (fromE. coli)
77682-594
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV
- Protein Synonyms:gp41 immunodominant region
- Protein/Peptide Name:gp41
- Purity:Greater than 90.0% as determined by SDS-page
- Molecular Weight:94 kDa
- Endotoxin Level:Very low
- Formulation:1.5 M urea, 25 mM tris-HCl pH 8.0 and 50% Glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-594
Recombinant HIV Type-O gp41 Antigen, Beta-galactosidase-tagged.