S. pneumoniae Recombinant Streptococcus pneumoniae type 22f Capsular Polysaccharide Antigen
Catalog # 77682-592
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:S. pneumoniae
- Protein/Peptide Name:Streptococcus pneumoniae type 22f Capsular Polysaccharide Antigen
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-592
Specifications
About this item
Recombinant Streptococcus pneumoniae type 22f Capsular Polysaccharide Antigen.