HeV Recombinant HEV
Catalog # 77682-590
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:HeV
- Protein Synonyms:Hepatitis E Virus
- Protein/Peptide Name:HEV
- Purity:Greater than 95.0% as determined by analysis by SDS-page
- Endotoxin Level:Very low
- Formulation:20 mM tris-HCl, pH-8, 10 mM B-ME and 8 M urea
- Shipping Temperature:Dry Ice
- Cat. No.:77682-590
Specifications
About this item
Recombinant HEV Antigen, b-galactosidase-tagged.