HSV-8 Recombinant ORF65
Catalog # 77682-580
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:HSV-8
- Protein Synonyms:HSV8|HSV-8 ORF65 and ORF8|ORF8|HSV-8 ORF65|HSV-8 ORF8
- Protein/Peptide Name:ORF65
- Purity:HSV-8 mosaic protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin Level:Very low
- Formulation:100 mM NaCl, 0.1% SDS and 50% glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-580
Specifications
About this item
Recombinant HSV-8 ORF65 and ORF8 Antigen, GST-tagged.