Hepatitis C virus (HCV) Recombinant NS5
77682-556
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:non structural protein 5|nonstructural antigen 5|nonstructural protein 5
- Protein/Peptide Name:NS5
- Purity:HCV NS5 genotype-3a protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin Level:Very low
- Formulation:1.5M urea, 25 mM tris-HCl, pH-8, 0.2% Triton-X and amp 50% Glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-556
Recombinant HCV Genotype-3a NS5 Antigen, GST-tagged.