Hepatitis C virus (HCV) Recombinant NS5
Catalog # 77682-548
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:non structural protein 5|nonstructural antigen 5|nonstructural protein 5
- Protein/Peptide Name:NS5
- Purity:HCV NS5 genotype-2 protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin Level:Very low
- Formulation:25 mM tris and 1 mM EDTA, 1.5M urea and amp 50% glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-548
Specifications
About this item
Recombinant HCV Genotype-2 NS5 Antigen.