Hepatitis C virus (HCV) Recombinant NS4
Catalog # 77682-524
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:nonstructural antigen 4|p8|Non structural protein 4A|p27|NS4B|NS4 a+b|NS4 immunodominant regions|Non structural protein 4B|NS4A
- Protein/Peptide Name:NS4
- Purity:HCV NS4 a+b fluoroscein protein is >95% pure as determined by 10% page (coomassie staining)
- Molecular Weight:19 kDa
- Endotoxin Level:Very low
- Formulation:20 mM tris-Hcl pH 8, 8 M urea and amp 10 mM B-ME
- Shipping Temperature:Dry Ice
- Cat. No.:77682-524
Specifications
About this item
Recombinant HCV NS4 a+b Antigen, b-galactosidase-tagged, Fluoroscein-Labled.