Hepatitis C virus (HCV) Recombinant NS3 immunodominant regions
77682-508
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:NS3P|NS3|nonstructural antigen 3|p70|Serine protease/NTPase/helicase|Non structural protein 3
- Protein/Peptide Name:NS3 immunodominant regions
- Purity:HCV NS3 genotype-6a protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin Level:Very low
- Formulation:1.5M urea, 25 mM tris-HCl pH-8, 0.2% Triton-X and amp 50% Glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-508
Recombinant HCV Genotype-6a NS3 Antigen, His-tagged.