Hepatitis C virus (HCV) Recombinant HCV (fromE. coli)
Catalog # 77682-466
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:Hepatitis C Virus
- Protein/Peptide Name:HCV
- Purity:Greater than 95.0% as determined by SDS-page
- Endotoxin Level:Very low
- Formulation:Both Conjugate protein (Core-NS3-NS4-NS5) and Capture protein (Core-NS3-NS4-NS5 + HCV core are in PBS
- Shipping Temperature:Dry Ice
- Cat. No.:77682-466
Specifications
About this item
Recombinant HCV Antigen.