Hepatitis C virus (HCV) Recombinant Capsid protein C (fromE. coli)
77682-436
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:HCV|Core protein p19|Hepatitis C virus|Hepatitis C virus core|HCV nucleocapsid|Core protein p21
- Protein/Peptide Name:Capsid protein C
- Purity:HCV Core genotype-1b protein is >95% pure as determined by SDS-page
- Endotoxin Level:Very low
- Formulation:(1 mg/ml) 1.5M urea, 25 mM tris-HCl, pH 8.0, 0.2% Triton-X and 50% glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-436
Recombinant HCV Core Nucleocapsid (Genotype-1b) Antigen, GST-tagged.