Hepatitis C virus (HCV) Recombinant Capsid protein C
77682-426
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein Synonyms:HCV|Core protein p19|Hepatitis C virus|Hepatitis C virus core|HCV nucleocapsid|Core protein p21
- Protein/Peptide Name:Capsid protein C
- Purity:HCV-Core Protein is >95% pure as determined by 10% page (coomassie staining)
- Molecular Weight:22 kDa
- Endotoxin Level:Very low
- Formulation:20 mM tris-Hcl pH 8, 8 M urea and 10 mM B-ME
- Shipping Temperature:Dry Ice
- Cat. No.:77682-426
Recombinant HCV Core nucleocapsid Antigen, b-galactosidase-tagged, FLSCN-Labled.