Influenza B Recombinant Hemagglutinin
Catalog # 77682-400
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Influenza B
- Protein Synonyms:Influenza B virus B/Singapore/INFTT-16-0610/2016 Hemagglutinin (MDCK cell derived, propiolactone inactivated)|Influenza B virus|Influenza virus type B
- Protein/Peptide Name:Hemagglutinin
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-400
Specifications
About this item
Recombinant Influenza B virus B/Singapore/INFTT-16-0610/2016 Hemagglutinin antigen (MDCK cell derived, propiolactone inactivated).