Hepatitis A virus (HAV) Recombinant VP1 immunodominant regions (fromE. coli)
Catalog # 77682-358
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Hepatitis A virus (HAV)
- Protein Synonyms:VP1
- Protein/Peptide Name:VP1 immunodominant regions
- Purity:HAV VP1 protein is >90% pure as determined by 10% page (coomassie staining)
- Molecular Weight:48 kDa
- Endotoxin Level:Very low
- Formulation:10 mM CBB, pH9.6, 0.1% SDS and 50% glycerol
- Shipping Temperature:Dry Ice
- Cat. No.:77682-358
Specifications
About this item
Recombinant HAV VP1 Antigen.