B. pertussis Bordetella pertussis Pertactin Antigen (formaldehyde inactivated)
Catalog # 77682-332
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Species:B. pertussis
- Protein/Peptide Name:Bordetella pertussis Pertactin Antigen (formaldehyde inactivated)
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-332
Specifications
About this item
Bordetella pertussis Pertactin Antigen (formaldehyde inactivated).