T. pallidum Recombinant p41 immunodominant regions (from E. coli)
Catalog # 77682-908
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:T. pallidum
- Protein Synonyms:p41 immunodominant regions|p41
- Protein/Peptide Name:p41 immunodominant regions
- Purity:Treponema pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining).
- Endotoxin Level:Very low
- Formulation:10 mM Tris-HCl pH8.0, 1 mM EDTA, 1 mMDTT and 8 M urea.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-908
Specifications
About this item
Recombinant T. pallidum p41 antigen, b-galactosidase-tagged.