T. pallidum Recombinant p15 immunodominant regions (from E. coli)
Catalog # 77682-900
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:T. pallidum
- Protein Synonyms:p15 immunodominant regions|Partial p15|p15
- Protein/Peptide Name:p15 immunodominant regions
- Purity:Treponema Pallidum protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Endotoxin Level:Very low
- Formulation:70 mM Tris-HCl pH-8, 84 mM NaCl, 14 mM Glutathione, 30% Glycerol & 0.2% Sarcosil.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-900
Specifications
About this item
Recombinant T. pallidum Partial p15 antigen, GST-tagged.