Toxoplasma gondii Recombinant p30 (SAG1) immunodominant regions (from E. coli)
Catalog # 77682-882
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Toxoplasma gondii
- Protein Synonyms:p30 (SAG1) immunodominant regions|SAG1|p30
- Protein/Peptide Name:p30 (SAG1) immunodominant regions
- Purity:Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Molecular Weight:42.3 kDa
- Endotoxin Level:Very low
- Formulation:50 mM Tris-HCl pH 8, 1.5 M urea and 50% glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-882
Specifications
About this item
Recombinant T. gondii p30 antigen, GST-tagged.