TBEV Recombinant glycoprotein E (from E. coli)
Vial
Catalog # 77682-868
Supplier: Creative Biomart
Supplier #: TBE-069
TBEV Recombinant glycoprotein E (from E. coli)
Catalog # 77682-868
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant Tick-Borne Encephalitis Virus gE antigen.
Specifications
Specifications
- Pack type:Vial
- Cat. No.:77682-868
- Shipping Temperature:Dry Ice
- Protein/Peptide Type:Recombinant
- Formulation:20 mM MES pH 6.5, 8 M urea, 200 mM NaCl and 0.05% Tween-20.
- Endotoxin Level:Very low
- Purity:Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Protein Synonyms:gE|glycoprotein E
- Protein/Peptide Name:glycoprotein E
- Species:TBEV
- Source:E. coli



