C. diphtheriae Corynebacterium diphtheriae
Catalog # 77682-856
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Species:Corynebacterium diphtheriae
- Protein Synonyms:Corynebacterium diphtheriae
- Protein/Peptide Name:Corynebacterium diphtheriae
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-856
Specifications
About this item
Corynebacterium diphtheriae.