HSV-2 gG protein
Catalog # 77682-850
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein Synonyms:gG|gG immunodominant regions|HSV-2 gG protein|HSV-2 gG immunodominant regions
- Protein/Peptide Name:HSV-2 gG protein
- Purity:Protein is >99% pure as determined by Analytical HPLC and Mass Spec.
- Endotoxin Level:Very low
- Formulation:HSV-2 gG solution contains 25 mM Tris-HCl pH8.0.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-850
Specifications
About this item
HSV-2 gG Protein.