HIV type-O Envelope
77682-848
New Product
:
- Pack type:Vial
- Protein Synonyms:HIV type-O Envelope|E|Envelope
- Protein/Peptide Name:HIV type-O Envelope
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Molecular Weight:2.6 kDa
- Endotoxin Level:Very low
- Formulation:1 mg/ml in water.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-848
HIV Type-O Envelope.