Order Entry
United States
ContactUsLinkComponent
SARS Recombinant surface glycoprotein (from E. coli)
SARS Recombinant surface glycoprotein (from E. coli)
Catalog # 77682-846
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
SARS Recombinant surface glycoprotein (from E. coli)
Catalog # 77682-846
Supplier:  Creative Biomart
Supplier Number:  SARS-411
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    SARS
  • Protein Synonyms:
    structural protein|Spike glycoprotein|surface glycoprotein|spike protein|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|S glycoprotein|E2 glycoprotein
  • Protein/Peptide Name:
    surface glycoprotein
  • Purity:
    Protein is >90% pure as determined SDS-PAGE.
  • Endotoxin Level:
    Very low
  • Formulation:
    SARS Spike protein solution is supplied in PBS.
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    77682-846

Specifications

About this item

Recombinant SARS Spike Protein, his-tagged.