Order Entry
United States
Orders LinkContactUsLinkComponent
HBcAg (from E. coli)

HBcAg (from E. coli)

Vial
Catalog # 77682-824
Supplier:   Creative Biomart
Supplier #:  SARS-400
HBcAg (from E. coli)
Catalog # 77682-824
Supplier:  Creative Biomart
CAS Number:  
HBcAg (from E. coli)
HBcAg (from E. coli)

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

HBcAg antigen.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-824
  • Molecular Weight:
    20 kDa
  • Shipping Temperature:
    Dry Ice
  • Formulation:
    Phosphate buffered saline, pH 7.4.
  • Endotoxin Level:
    Very low
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Protein Synonyms:
    HBcAg adw2|adw2|HBcAg
  • Protein/Peptide Name:
    HBcAg
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.