Recombinant Havcr1 (from Sf9, Baculovirus cells)
Catalog # 77682-822
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Sf9 Insect cells)
- Protein Synonyms:TIM-1|KIM-1|AI503787|t cell immunoglobulin and mucin domain-containing protein 1|Havcr1|T-cell immunoglobulin mucin receptor 1|Timd1|hepatitis A virus cellular receptor 1|T-cell immunoglobulin and mucin domain containing 1|hepatitis A virus cellular receptor 1 homolog|t cell membrane protein 1|kidney injury molecule 1|HAVcr-1|TIMD-1|Tim1
- Protein/Peptide Name:Havcr1
- Purity:Greater than 90.0% as determined by SDS-PAGE.
- Molecular Weight:24.4 kDa
- Endotoxin Level:Very low
- Formulation:HAVCR1 protein solution (0.25 mg/ml) contains PhosphateBuffered Saline (pH 7.4) and 10% glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-822
Specifications
About this item
Recombinant Mouse Havcr1 Protein, his-tagged.