HIV-1 Recombinant HIV-1 gag p17 and p24 (from E. coli)
77682-798
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV-1
- Protein Synonyms:p24|HIV-1 gag|HIV-1 gag p17 and p24|p17
- Protein/Peptide Name:HIV-1 gag p17 and p24
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Endotoxin Level:Very low
- Formulation:1.5 M urea, 25 mM Tris-HCl pH 8.0, 0.2% Triton-X & 50% Glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-798
Recombinant HIV-1 gag p17 and p24 antigen, GST-tagged.