Mumps virus Recombinant Nucleoprotein (from E. coli)
Catalog # 77682-766
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mumps virus
- Protein Synonyms:Nucleoprotein|NP
- Protein/Peptide Name:Nucleoprotein
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining).
- Molecular Weight:62 kDa
- Endotoxin Level:Very low
- Formulation:Phosphate buffer, pH 7.4.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-766
Specifications
About this item
Recombinant Mumps viral Nucleoprotein antigen, his-tagged.