Ebolavirus Recombinant Nucleoprotein (from E. coli)
Catalog # 77682-752
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Ebolavirus
- Protein Synonyms:Nucleoprotein|NP
- Protein/Peptide Name:Nucleoprotein
- Purity:>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain).
- Molecular Weight:15 kDa
- Endotoxin Level:Very low
- Formulation:Ebola Sudan protein solution is supplied in Phosphate buffer and 0.02% sodium azide.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-752
Specifications
About this item
Recombinant Ebola Sudan Nucleoprotein antigen.