HTLV-1 Recombinant gp21 and gp46 immunodominant regions
Catalog # 77682-728
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:HTLV-1
- Protein Synonyms:gp46 mosaic|gp21 mosaic|gp46 immunodominant region|gp21|gp21 immunodominant region|gp46|gp21 and gp46 immunodominant regions
- Protein/Peptide Name:gp21 and gp46 immunodominant regions
- Purity:HTLV-1 Mosaic protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
- Molecular Weight:42 kDa
- Endotoxin Level:Very low
- Formulation:60 mM NaCl, 50 mM Tris-HCl pH 8.0, 0.25% Sarkosil, 50% glycerol and 10 mM glutathione.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-728
Specifications
About this item
Recombinant HTLV-1 gp21 and gp46 mosaic antigen, GST-tagged.